Assay Details
| Gene Symbol: SLC8A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293097.2 | NP_001280026.1 | 1 | 990 | 115 | |
| XM_015287282.2 | XP_015142768.1 | 3 | 7780 | 115 | ||
| XM_025150587.1 | XP_025006355.1 | 3 | 1385 | 115 | ||
| XM_025150588.1 | XP_025006356.1 | 3 | 7780 | 115 | ||
| GenBank mRNA | AJ012580.1 | - | 1 | 654 | 115 | |
More Information
Set Membership: |
|

SLC8A3
Genomic Map