Assay Details
| Gene Symbol: HIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277483.1 | NP_001264412.1 | 1 - 2 | 43 | 64 | |
| GenBank mRNA | BX929888.1 | - | 1 - 2 | 51 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277483.1 transcripts.
Set Membership: |
|

HIC2
Genomic Map