Assay Details
| Gene Symbol: QPCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149300.1 | XP_025005068.1 | 3 - 4 | 2061 | 57 | |
| XM_025149301.1 | XP_025005069.1 | 4 - 5 | 1558 | 57 | ||
| XM_419527.6 | XP_419527.3 | 3 - 4 | 1425 | 57 | ||
| GenBank mRNA | BX929950.1 | - | 1 - 2 | 144 | 57 | |
More Information
Set Membership: |
|

QPCT
Genomic Map