Assay Details
| Gene Symbol: LAMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015283531.2 | XP_015139017.1 | 4 - 5 | 654 | 59 | |
| GenBank mRNA | BX933313.1 | - | 4 - 5 | 462 | 59 | |
More Information
Set Membership: |
|

LAMP5
Genomic Map