Assay Details
| Gene Symbol: EMSY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025146941.1 | XP_025002709.1 | 18 - 19 | 2969 | 61 | |
| GenBank mRNA | BX934563.1 | - | 4 - 5 | 616 | 61 | |
More Information
Set Membership: |
|

EMSY
Genomic Map