Assay Details
| Gene Symbol: RPP38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199590.1 | NP_001186519.1 | 2 | 1025 | 74 | |
| XM_015281388.2 | XP_015136874.1 | 3 | 929 | 74 | ||
| XM_025147186.1 | XP_025002954.1 | 3 | 951 | 74 | ||
| GenBank mRNA | BX934704.1 | - | 1 | 545 | 74 | |
| CR406776.1 | - | 2 | 808 | 74 | ||
More Information
Set Membership: |
|

RPP38
Genomic Map