Assay Details
| Gene Symbol: ANXA8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288444.2 | XP_015143930.1 | 11 - 12 | 999 | 60 | |
| XM_421646.6 | XP_421646.1 | 11 - 12 | 905 | 60 | ||
| GenBank mRNA | BX935521.1 | - | 12 - 13 | 907 | 60 | |
More Information
Set Membership: |
|

ANXA8L1
Genomic Map