Assay Details
| Gene Symbol: HOXA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305224.1 | NP_001292153.1 | 1 - 2 | 584 | 62 | |
| GenBank mRNA | CR391427.1 | - | 1 - 2 | 584 | 62 | |
| X97750.1 | - | 266 | 62 | |||
More Information
Set Membership: |
|

HOXA9
Genomic Map