Assay Details
| Gene Symbol: PRDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641038.4 | XP_003641086.2 | 4 - 5 | 913 | 61 | |
| XM_004940353.3 | XP_004940410.1 | 4 - 5 | 782 | 61 | ||
| XM_004940355.3 | XP_004940412.1 | 6 - 7 | 1173 | 61 | ||
| XM_015284537.2 | XP_015140023.1 | 5 - 6 | 1100 | 61 | ||
| XM_015284538.2 | XP_015140024.1 | 6 - 7 | 969 | 61 | ||
| XM_015284539.2 | XP_015140025.1 | 2 - 3 | 585 | 61 | ||
| XM_025149056.1 | XP_025004824.1 | 4 - 5 | 755 | 61 | ||
| XM_025149057.1 | XP_025004825.1 | 4 - 5 | 1043 | 61 | ||
| XM_025149059.1 | XP_025004827.1 | 4 - 5 | 1293 | 61 | ||
| XM_025149060.1 | XP_025004828.1 | 4 - 5 | 643 | 61 | ||
| GenBank mRNA | AB278131.1 | - | 3 - 4 | 555 | 61 | |
More Information
Set Membership: |
|

PRDM1
Genomic Map