Assay Details
| Gene Symbol: POLDIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317356.1 | NP_001304285.1 | 2 - 3 | 286 | 63 | |
| GenBank mRNA | AJ719677.1 | - | 2 - 3 | 277 | 63 | |
More Information
Set Membership: |
|

POLDIP2
Genomic Map