Assay Details
| Gene Symbol: CZH9ORF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004949224.3 | XP_004949281.1 | 9 - 10 | 2097 | 104 | |
| XM_004949225.3 | XP_004949282.1 | 9 - 10 | 2097 | 104 | ||
| XM_025145254.1 | XP_025001022.1 | 9 - 10 | 2097 | 104 | ||
| XM_025145255.1 | XP_025001023.1 | 9 - 10 | 2057 | 104 | ||
| XM_025145256.1 | XP_025001024.1 | 9 - 10 | 2069 | 104 | ||
| GenBank mRNA | BX931259.2 | - | 6 - 7 | 700 | 104 | |
More Information
Set Membership: |
|

CZH9ORF3
Genomic Map