Assay Details
| Gene Symbol: CZH9ORF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004949224.3 | XP_004949281.1 | 11 - 12 | 2208 | 147 | |
| XM_004949225.3 | XP_004949282.1 | 11 - 12 | 2208 | 147 | ||
| XM_025145254.1 | XP_025001022.1 | 11 - 12 | 2208 | 147 | ||
| XM_025145255.1 | XP_025001023.1 | 11 - 12 | 2168 | 147 | ||
| XM_025145256.1 | XP_025001024.1 | 11 - 12 | 2180 | 147 | ||
| GenBank mRNA | BX931259.2 | - | 8 - 9 | 811 | 147 | |
More Information
Set Membership: |
|

CZH9ORF3
Genomic Map