Assay Details
| Gene Symbol: AMZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414779.5 | XP_414779.2 | 7 | 3133 | 126 | |
| GenBank mRNA | CR389729.1 | - | 1 | 452 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AMZ2| Gene Symbol: AMZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414779.5 | XP_414779.2 | 7 | 3133 | 126 | |
| GenBank mRNA | CR389729.1 | - | 1 | 452 | 126 | |
Set Membership: |
|