Assay Details
| Gene Symbol: ZNF532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642897.4 | XP_003642945.1 | 2 | 1079 | 149 | |
| XM_004949005.3 | XP_004949062.1 | 3 | 1207 | 149 | ||
| XM_004949006.3 | XP_004949063.1 | 2 | 1058 | 149 | ||
| XM_015277394.2 | XP_015132880.1 | 3 | 1313 | 149 | ||
| XM_015277395.2 | XP_015132881.1 | 3 | 1158 | 149 | ||
| XM_015277396.2 | XP_015132882.1 | 3 | 1196 | 149 | ||
| XM_015277397.2 | XP_015132883.1 | 3 | 1192 | 149 | ||
| XM_015277398.2 | XP_015132884.1 | 2 | 1057 | 149 | ||
| XM_015277399.2 | XP_015132885.1 | 3 | 1207 | 149 | ||
| GenBank mRNA | CR390053.1 | - | 1 | 249 | 149 | |
More Information
Set Membership: |
|

ZNF532
Genomic Map