Assay Details
| Gene Symbol: SNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419196.6 | XP_419196.2 | 1 - 2 | 319 | 94 | |
| GenBank mRNA | CR407272.1 | - | 1 - 2 | 174 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNAI2| Gene Symbol: SNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_419196.6 | XP_419196.2 | 1 - 2 | 319 | 94 | |
| GenBank mRNA | CR407272.1 | - | 1 - 2 | 174 | 94 | |
Set Membership: |
|