Assay Details
| Gene Symbol: PUS7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937548.3 | XP_004937605.1 | 2 | 767 | 153 | |
| XM_015282485.2 | XP_015137971.1 | 2 | 721 | 153 | ||
| XM_416038.6 | XP_416038.1 | 2 | 741 | 153 | ||
| GenBank mRNA | BX930408.2 | - | 2 | 656 | 153 | |
More Information
Set Membership: |
|

PUS7L
Genomic Map