Assay Details
| Gene Symbol: MRPL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277471.1 | NP_001264400.1 | 7 | 747 | 119 | |
| XM_025147134.1 | XP_025002902.1 | 7 | 777 | 119 | ||
| GenBank mRNA | AJ442248.1 | - | 2 | 294 | 119 | |
| BU339965.1 | - | 3 | 384 | 119 | ||
| BX933852.2 | - | 7 | 726 | 119 | ||
| BX934702.1 | - | 7 | 743 | 119 | ||
| CV861166.1 | - | 7 | 722 | 119 | ||
More Information
Set Membership: |
|

MRPL13
Genomic Map