Assay Details
| Gene Symbol: METTL2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006329.1 | NP_001006329.1 | 2 - 3 | 203 | 136 | |
| GenBank mRNA | AJ721086.1 | - | 2 - 3 | 203 | 136 | |
| BX931317.2 | - | 2 - 3 | 163 | 136 | ||
More Information
Set Membership: |
|

METTL2B
Genomic Map