Assay Details
| Gene Symbol: METTL2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006329.1 | NP_001006329.1 | 5 - 6 | 672 | 102 | |
| GenBank mRNA | AJ721086.1 | - | 5 - 6 | 672 | 102 | |
| BX931317.2 | - | 6 - 7 | 455 | 102 | ||
| CR354048.1 | - | 3 - 4 | 397 | 102 | ||
| CR386397.1 | - | 4 - 5 | 542 | 102 | ||
More Information
Set Membership: |
|

METTL2B
Genomic Map