Assay Details
| Gene Symbol: RNF126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006338.2 | NP_001006338.2 | 3 - 4 | 211 | 75 | |
| XM_015299919.2 | XP_015155405.1 | 3 - 4 | 75 | |||
| GenBank mRNA | AJ720373.1 | - | 3 - 4 | 211 | 75 | |
More Information
Set Membership: |
|

RNF126
Genomic Map