Assay Details
| Gene Symbol: EIF3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006349.1 | NP_001006349.1 | 5 - 6 | 487 | 75 | |
| GenBank mRNA | AJ719829.1 | - | 5 - 6 | 487 | 75 | |
More Information
Set Membership: |
|

EIF3E
Genomic Map