Assay Details
| Gene Symbol: CEPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006392.2 | NP_001006392.1 | 1031 | 92 | ||
| XM_015298707.2 | XP_015154193.1 | 1079 | 92 | |||
| XM_015298708.2 | XP_015154194.1 | 1079 | 92 | |||
| XM_015298709.2 | XP_015154195.1 | 951 | 92 | |||
| XM_015298710.2 | XP_015154196.1 | 1311 | 92 | |||
| XM_015298711.2 | XP_015154197.1 | 1311 | 92 | |||
| XM_025143711.1 | XP_024999479.1 | 975 | 92 | |||
| GenBank mRNA | AJ720153.1 | - | 1033 | 92 | ||
More Information
Set Membership: |
|

CEPT1
Genomic Map