Assay Details
| Gene Symbol: ATG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006409.1 | NP_001006409.1 | 4 - 5 | 413 | 66 | |
| XM_015284516.2 | XP_015140002.1 | 4 - 5 | 385 | 66 | ||
| XM_015284517.2 | XP_015140003.1 | 4 - 5 | 490 | 66 | ||
| XM_015284518.2 | XP_015140004.1 | 4 - 5 | 633 | 66 | ||
| XM_015284519.2 | XP_015140005.1 | 4 - 5 | 416 | 66 | ||
| XM_015284520.2 | XP_015140006.1 | 4 - 5 | 385 | 66 | ||
| XM_015284521.2 | XP_015140007.1 | 4 - 5 | 385 | 66 | ||
| XM_015284523.2 | XP_015140009.1 | 4 - 5 | 385 | 66 | ||
| GenBank mRNA | AJ721022.1 | - | 4 - 5 | 413 | 66 | |
More Information
Set Membership: |
|

ATG5
Genomic Map