Assay Details
| Gene Symbol: HN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006425.1 | NP_001006425.1 | 3 - 4 | 422 | 76 | |
| XM_015295400.2 | XP_015150886.1 | 3 - 4 | 553 | 76 | ||
| GenBank mRNA | AJ719480.1 | - | 3 - 4 | 422 | 76 | |
More Information
Set Membership: |
|

HN1
Genomic Map