Assay Details
| Gene Symbol: SLC25A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006443.1 | NP_001006443.1 | 2 - 3 | 693 | 104 | |
| GenBank mRNA | AJ719388.1 | - | 2 - 3 | 693 | 104 | |
More Information
Set Membership: |
|

SLC25A4
Genomic Map