Assay Details
| Gene Symbol: EIF2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006477.1 | NP_001006477.1 | 3 - 4 | 422 | 71 | |
| XM_025150723.1 | XP_025006491.1 | 3 - 4 | 416 | 71 | ||
| GenBank mRNA | AJ719612.1 | - | 3 - 4 | 422 | 71 | |
More Information
Set Membership: |
|

EIF2S1
Genomic Map