Assay Details
| Gene Symbol: SULT1C3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204601.2 | NP_989932.1 | 7 - 8 | 865 | 72 | |
| XM_015277822.2 | XP_015133308.1 | 8 - 9 | 973 | 72 | ||
| GenBank mRNA | AJ416889.1 | - | 7 - 8 | 866 | 72 | |
More Information
Set Membership: |
|

SULT1C3
Genomic Map