Assay Details
| Gene Symbol: CENPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204611.1 | NP_989942.1 | 8 - 9 | 636 | 75 | |
| GenBank mRNA | AB055783.1 | - | 8 - 9 | 636 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CENPH| Gene Symbol: CENPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204611.1 | NP_989942.1 | 8 - 9 | 636 | 75 | |
| GenBank mRNA | AB055783.1 | - | 8 - 9 | 636 | 75 | |
Set Membership: |
|