Assay Details
| Gene Symbol: CNTN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204612.1 | NP_989943.1 | 18 - 19 | 2451 | 67 | |
| XM_015279870.2 | XP_015135356.1 | 18 - 19 | 2599 | 67 | ||
| XM_015279875.2 | XP_015135361.1 | 18 - 19 | 3195 | 67 | ||
| XM_015279880.2 | XP_015135366.1 | 17 - 18 | 2459 | 67 | ||
| XM_015279889.2 | XP_015135375.1 | 16 - 17 | 2792 | 67 | ||
| GenBank mRNA | AJ309935.1 | - | 18 - 19 | 2451 | 67 | |
More Information
Set Membership: |
|

CNTN5
Genomic Map