Assay Details
| Gene Symbol: PSMA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204613.1 | NP_989944.1 | 2 - 3 | 246 | 86 | |
| GenBank mRNA | CR387264.1 | - | 2 - 3 | 231 | 86 | |
| U09226.1 | - | 2 - 3 | 246 | 86 | ||
More Information
Set Membership: |
|

PSMA7
Genomic Map