Assay Details
Gene Symbol: OPN4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204625.3 | NP_989956.3 | 7 - 8 | 1097 | 101 | |
XM_015276115.2 | XP_015131601.1 | 17 - 18 | 3923 | 101 | ||
XM_015276116.2 | XP_015131602.1 | 17 - 18 | 8887 | 101 | ||
XM_015276120.2 | XP_015131606.1 | 9 - 10 | 1976 | 101 | ||
XM_025149493.1 | XP_025005261.1 | 17 - 18 | 8961 | 101 | ||
XM_025149494.1 | XP_025005262.1 | 15 - 16 | 8991 | 101 | ||
XM_025149495.1 | XP_025005263.1 | 17 - 18 | 6099 | 101 | ||
XM_025149496.1 | XP_025005264.1 | 16 - 17 | 8965 | 101 | ||
XM_025149497.1 | XP_025005265.1 | 17 - 18 | 8952 | 101 | ||
XM_025149498.1 | XP_025005266.1 | 16 - 17 | 8777 | 101 | ||
GenBank mRNA | AY036061.1 | - | 7 - 8 | 1099 | 101 | |
EU124630.1 | - | 7 - 8 | 1099 | 101 | ||
EU124631.1 | - | 7 - 8 | 1099 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
