Assay Details
| Gene Symbol: MXRA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204636.2 | NP_989967.1 | 8 - 9 | 1255 | 77 | |
| XM_015296820.2 | XP_015152306.1 | 9 - 10 | 1416 | 77 | ||
| XM_025142483.1 | XP_024998251.1 | 10 - 11 | 1464 | 77 | ||
| GenBank mRNA | AF373843.1 | - | 8 - 9 | 1259 | 77 | |
More Information
Set Membership: |
|

MXRA8
Genomic Map