Assay Details
| Gene Symbol: AvBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204650.2 | NP_989981.1 | 1 - 2 | 60 | 83 | |
| GenBank mRNA | AF181952.1 | - | 1 - 2 | 63 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AvBD3| Gene Symbol: AvBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204650.2 | NP_989981.1 | 1 - 2 | 60 | 83 | |
| GenBank mRNA | AF181952.1 | - | 1 - 2 | 63 | 83 | |
Set Membership: |
|