Assay Details
| Gene Symbol: AvBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204650.2 | NP_989981.1 | 2 - 3 | 171 | 89 | |
| GenBank mRNA | AF181952.1 | - | 2 - 3 | 174 | 89 | |
| DQ677634.1 | - | 2 - 3 | 145 | 89 | ||
| DQ858300.1 | - | 2 - 3 | 145 | 89 | ||
| DQ858313.1 | - | 2 - 3 | 145 | 89 | ||
| DQ858326.1 | - | 2 - 3 | 145 | 89 | ||
| DQ858340.1 | - | 2 - 3 | 145 | 89 | ||
More Information
Set Membership: |
|

AvBD3
Genomic Map