Assay Details
| Gene Symbol: GPR149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204662.1 | NP_989993.1 | 3 - 4 | 2115 | 75 | |
| XM_015291641.2 | XP_015147127.1 | 3 - 4 | 5063 | 75 | ||
| XM_015291642.2 | XP_015147128.1 | 6 - 7 | 4770 | 75 | ||
| XM_015291643.2 | XP_015147129.1 | 6 - 7 | 1376 | 75 | ||
| XM_015291644.2 | XP_015147130.1 | 6 - 7 | 1513 | 75 | ||
| XM_015291646.2 | XP_015147132.1 | 7 - 8 | 4850 | 75 | ||
| XM_015291648.2 | XP_015147134.1 | 7 - 8 | 4853 | 75 | ||
| XM_025153308.1 | XP_025009076.1 | 5 - 6 | 1266 | 75 | ||
| GenBank mRNA | AJ293971.1 | - | 3 - 4 | 2115 | 75 | |
More Information
Set Membership: |
|

GPR149
Genomic Map