Assay Details
| Gene Symbol: FGF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204674.2 | NP_990005.2 | 1 - 2 | 365 | 96 | |
| GenBank mRNA | AF315355.1 | - | 1 - 2 | 374 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGF19| Gene Symbol: FGF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204674.2 | NP_990005.2 | 1 - 2 | 365 | 96 | |
| GenBank mRNA | AF315355.1 | - | 1 - 2 | 374 | 96 | |
Set Membership: |
|