Assay Details
| Gene Symbol: FGF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204674.2 | NP_990005.2 | 2 - 3 | 466 | 94 | |
| GenBank mRNA | AB111892.1 | - | 1 - 2 | 109 | 94 | |
| AF315355.1 | - | 2 - 3 | 475 | 94 | ||
More Information
Set Membership: |
|

FGF19
Genomic Map