Assay Details
| Gene Symbol: CDX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204676.2 | NP_990007.2 | 1 - 2 | 515 | 69 | |
| GenBank mRNA | AB046532.1 | - | 1 - 2 | 515 | 69 | |
| HG313857.1 | - | 1 - 2 | 321 | 69 | ||
More Information
Set Membership: |
|

CDX1
Genomic Map