Assay Details
| Gene Symbol: TRA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204678.1 | NP_990009.1 | 3 - 4 | 422 | 63 | |
| XM_015276811.2 | XP_015132297.1 | 3 - 4 | 446 | 63 | ||
| GenBank mRNA | AF282244.1 | - | 3 - 4 | 422 | 63 | |
| BX934662.1 | - | 3 - 4 | 368 | 63 | ||
More Information
Set Membership: |
|

TRA2B
Genomic Map