Assay Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204686.2 | NP_990017.1 | 1 - 2 | 299 | 81 | |
| XM_025142764.1 | XP_024998532.1 | 4 - 5 | 2827 | 81 | ||
| GenBank mRNA | AF220436.1 | - | 1 - 2 | 302 | 81 | |
More Information
Set Membership: |
|

STAR
Genomic Map