Assay Details
| Gene Symbol: CCL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204720.1 | NP_990051.1 | 1 - 2 | 127 | 122 | |
| GenBank mRNA | MK284230.1 | - | 1 - 2 | 103 | 122 | |
| Y18692.1 | - | 127 | 122 | |||
More Information
Set Membership: |
|

CCL4
Genomic Map