Assay Details
| Gene Symbol: CHRNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204814.1 | NP_990145.1 | 5 - 6 | 1751 | 59 | |
| GenBank mRNA | AJ250361.1 | - | 5 - 6 | 1751 | 59 | |
More Information
Set Membership: |
|

CHRNA4
Genomic Map