Assay Details
| Gene Symbol: CHRNB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204819.1 | NP_990150.1 | 5 - 6 | 1478 | 66 | |
| GenBank mRNA | J05643.2 | - | 5 - 6 | 1478 | 66 | |
More Information
Set Membership: |
|

CHRNB4
Genomic Map