Assay Details
| Gene Symbol: EFNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204824.1 | NP_990155.1 | 4 - 5 | 616 | 61 | |
| GenBank mRNA | AF180729.1 | - | 4 - 5 | 616 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EFNB2| Gene Symbol: EFNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204824.1 | NP_990155.1 | 4 - 5 | 616 | 61 | |
| GenBank mRNA | AF180729.1 | - | 4 - 5 | 616 | 61 | |
Set Membership: |
|