Assay Details
| Gene Symbol: ELAVL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204833.1 | NP_990164.1 | 2 - 3 | 248 | 83 | |
| XM_015299667.2 | XP_015155153.1 | 2 - 3 | 252 | 83 | ||
| XM_015299669.2 | XP_015155155.1 | 2 - 3 | 204 | 83 | ||
| XM_015299670.2 | XP_015155156.1 | 2 - 3 | 248 | 83 | ||
| XM_025144245.1 | XP_025000013.1 | 2 - 3 | 251 | 83 | ||
| GenBank mRNA | AF176673.1 | - | 2 - 3 | 248 | 83 | |
More Information
Set Membership: |
|

ELAVL1
Genomic Map