Assay Details
| Gene Symbol: DUSP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204838.1 | NP_990169.1 | 2 - 3 | 550 | 80 | |
| GenBank mRNA | AF167296.1 | - | 2 - 3 | 550 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUSP4| Gene Symbol: DUSP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204838.1 | NP_990169.1 | 2 - 3 | 550 | 80 | |
| GenBank mRNA | AF167296.1 | - | 2 - 3 | 550 | 80 | |
Set Membership: |
|