Assay Details
| Gene Symbol: WSB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204872.1 | NP_990203.1 | 6 - 7 | 962 | 85 | |
| GenBank mRNA | AF072879.1 | - | 6 - 7 | 962 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WSB1| Gene Symbol: WSB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204872.1 | NP_990203.1 | 6 - 7 | 962 | 85 | |
| GenBank mRNA | AF072879.1 | - | 6 - 7 | 962 | 85 | |
Set Membership: |
|