Assay Details
| Gene Symbol: WSB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204872.1 | NP_990203.1 | 7 - 8 | 1084 | 125 | |
| GenBank mRNA | AF072879.1 | - | 7 - 8 | 1084 | 125 | |
More Information
Set Membership: |
|

WSB1
Genomic Map