Assay Details
| Gene Symbol: PTPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204875.1 | NP_990206.1 | 3 - 4 | 370 | 73 | |
| XM_015296330.2 | XP_015151816.1 | 3 - 4 | 475 | 73 | ||
| XM_015296332.2 | XP_015151818.1 | 4 - 5 | 550 | 73 | ||
| XM_015296333.2 | XP_015151819.1 | 5 - 6 | 692 | 73 | ||
| XM_025142161.1 | XP_024997929.1 | 6 - 7 | 826 | 73 | ||
| XM_025142162.1 | XP_024997930.1 | 3 - 4 | 475 | 73 | ||
| GenBank mRNA | L20630.1 | - | 1 - 2 | 74 | 73 | |
| U46662.1 | - | 3 - 4 | 222 | 73 | ||
| U86410.1 | - | 3 - 4 | 370 | 73 | ||
More Information
Set Membership: |
|

PTPN1
Genomic Map