Assay Details
| Gene Symbol: EZR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204885.1 | NP_990216.1 | 11 - 12 | 1357 | 76 | |
| XM_015284013.2 | XP_015139499.1 | 8 - 9 | 938 | 76 | ||
| GenBank mRNA | AB019790.1 | - | 11 - 12 | 1357 | 76 | |
More Information
Set Membership: |
|

EZR
Genomic Map